Order Entry
Austria
ContactUsLinkComponent
CLP Protocol Test Kit for ICP Analysis, Specpure®
CLP Protocol Test Kit for ICP Analysis, Specpure®
Certificates
CLP Protocol Test Kit for ICP Analysis, Specpure®
Certificates

Specifications

  • MDL No.:
    MFCD00147250
  • UN No.:
    3264
  • Physical form:
    Liquid
  • Molecular formula:
    Matrix: 5% HCl/1% HNO3

Specifications

Frequently Bought Together