Order Entry
Austria
ContactUsLinkComponent
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 40 mesh woven from 0.25mm (0.01in) Type 304
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 40 mesh woven from 0.25mm (0.01in) Type 304
Catalog # 13478.RR
CAS Number:  
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 40 mesh woven from 0.25mm (0.01in) Type 304
Catalog # 13478.RR
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    7615
  • Environmentally Preferable:
  • Pk Info:
    10×10 cm

Specifications

About this item

 :
 :55.845 g/mol
Storage Temperature:Ambient
Formula:Fe
MDL Number:MFCD00148499