Calcium zirconium oxide ≥99.2% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12013-47-7
:
Formula:CaZrO₃
MDL Number:MFCD00015982
:179.30 g/mol
Melting Pt:2550 °C
Storage Temperature:Ambient
EINECS:234-586-6
Related Information
Frequently Bought Together