Order Entry
Austria
ContactUsLinkComponent
 
VWR® VisiScope® ALC, Upright Microscopes
 
undefined
VWR® VisiScope® ALC, Upright Microscopes

 

  • Catalog No:
  • 630-1925
  • 630-1927
  • Observation method:
  • Brightfield
  • Brightfield
  • Eyepieces:
  • WF 10×/18 mm
  • WF 10×/18 mm
  • Nosepiece:
  • Quadruple
  • Quadruple
  • Objectives:
  • Achromatic HC 4×, 10×, 40×, 100× (i)
  • Achromatic HC 4×, 10×, 40×, 100× (i)
  • Stage:
  • 125×116 mm (70×30 mm movement)
  • 125×116 mm (70×30 mm movement)
  • Condenser:
  • Abbe N.A. 1.25, pre-centered, focusable, with iris diaphragm
  • Abbe N.A. 1.25, pre-centered, focusable, with iris diaphragm
  • Focusing system:
  • Coaxial coarse and fine knobs
  • Coaxial coarse and fine knobs
  • Illumination:
  • LED (1 W)
  • LED (1 W)
  • W×D×H:
  • 170×210×350 mm
  • 170×210×350 mm
  • Weight:
  • 4 kg
  • 4 kg

 

Eyepieces

 

Frequently Bought Together