Anti-CTNNB1 Goat Polyclonal Antibody
Catalog # EVBIEB12543
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:catenin (cadherin-associated protein), beta 1, 88kDa
- Antigen symbol:CTNNB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Pig,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Format:affinity purified
- Form:liquid
- Gene ID:1499
- Antigen synonyms:armadillo|MRD19|CTNNB
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Sequence:C-QIPSTQFDAAHPTN
- Storage temperature:Store at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing
- Concentration:100µg/200µl
- Shipping temperature:8 °C
- Purification:Affinity purified
- Size:100µg
- Pack type:plastic vial
- Pk:0,1 mg
Specifications
About this item
Western Blot: Approx 100kDa band observed in lysates of cell line HeLa (calculated MW of 85.5kDa according to NP_001895.1). Recommended concentration: 0.3-1µg/ml.
Type: Primary
Antigen: CTNNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Pig, Bovine