Orders LinkContactUsLinkComponent
Vials
Vials
Vial with 2, 5, 10, 15, 20 and 25 ml graduations marks, for LeadTrak™, Lead DR 100, FF3, IR24, BC3, SADT, 25,00 ml, Yes
Catalog # HACH219300
Supplier:   Hach
Vials
Catalog # HACH219300
Supplier:  Hach
Vials
Vials
Vial with 2, 5, 10, 15, 20 and 25 ml graduations marks, for LeadTrak™, Lead DR 100, FF3, IR24, BC3, SADT, 25,00 ml, Yes
Price
9,50 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Vial with 2, 5, 10, 15, 20 and 25 ml marks, for LeadTrak™, Lead DR 100, FF3, IR24, BC3, SADT.
Specifications
  • Cap included:
    Yes
  • Capacity:
    25,00 ml
  • Description:
    Vial with 2, 5, 10, 15, 20 and 25 ml graduations marks, for LeadTrak™, Lead DR 100, FF3, IR24, BC3, SADT