Orders LinkContactUsLinkComponent
Electrode filling solution
Electrode filling solution
Internal filling solution for IntelliCAL™ PHC301 pH probes and MTC301 ORP probes, 3,0M KCl, saturated with silver chloride (AgCl), 28 ml bottle, 28 mL
Catalog # HACH2841700
Supplier:   Hach
Electrode filling solution
Catalog # HACH2841700
Supplier:  Hach
CAS Number:  
Electrode filling solution
Electrode filling solution
Internal filling solution for IntelliCAL™ PHC301 pH probes and MTC301 ORP probes, 3,0M KCl, saturated with silver chloride (AgCl), 28 ml bottle, 28 mL
CAS Number:  
Price
27,30 €
Log in for availability
1 * 28 mL
Product Details & Documents
Specifications
Specifications
  • Pk:
    28 mL
  • Description:
    Internal filling solution for IntelliCAL™ PHC301 pH probes and MTC301 ORP probes, 3,0M KCl, saturated with silver chloride (AgCl), 28 ml bottle