Order Entry
Austria
Orders LinkContactUsLinkComponent
Electrode cleaning solution

Electrode cleaning solution

Electrode cleaning solution, for cleaning all pH, ORP and sodium ion selective eletrodes, 500 ml bottle, 500 mL
Catalog # 705-1031
Supplier:   Hach
Electrode cleaning solution
Catalog # 705-1031
Supplier:  Hach
CAS Number:  
Electrode cleaning solution
Electrode cleaning solution

Electrode cleaning solution, for cleaning all pH, ORP and sodium ion selective eletrodes, 500 ml bottle, 500 mL

CAS Number:  
Price
56,10 €
Log in for availability
1 * 500 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Electrode cleaning solution, for cleaning all pH, ORP and sodium ion selective eletrodes, 500 ml bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.