ContactUsLinkComponent
Multi-parameter meters, handheld, MultiLine® Multi 3510 IDS
Multi-parameter meters, handheld, MultiLine® Multi 3510 IDS
Supplier:  WTW
Certificates
undefined
Multi-parameter meters, handheld, MultiLine® Multi 3510 IDS
Supplier:  WTW
Specifications
  • Catalog No:
  • 662-2138
  • 662-2139
  • 662-2136
  • 662-2137
  • 662-2135
  • Model:
  • Multi 3510 IDS
  • Multi 3510 IDS
  • Multi 3510 IDS
  • Multi 3510 IDS
  • Multi 3510 IDS
  • Display:
  • Graphic LCD, backlit
  • Graphic LCD, backlit
  • Graphic LCD, backlit
  • Graphic LCD, backlit
  • Graphic LCD, backlit
  • Power supply:
  • 4× 1,5 V AA (supplied); 4× 1,2 V NiMH-Akku or via USB port
  • 4× 1,5 V AA (supplied); 4× 1,2 V NiMH-Akku or via USB port
  • 4× 1,5 V AA (supplied); 4× 1,2 V NiMH-Akku or via USB port
  • 4× 1,5 V AA (supplied); 4× 1,2 V NiMH-Akku or via USB port
  • 4× 1,5 V AA (supplied); 4× 1,2 V NiMH-Akku or via USB port
  • Compliance:
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • Data logging capacity:
  • 500 (manual); 4500 (automatic)
  • 500 (manual); 4500 (automatic)
  • 500 (manual); 4500 (automatic)
  • 500 (manual); 4500 (automatic)
  • 500 (manual); 4500 (automatic)
  • Connections:
  • 1 measurement input
  • 1 measurement input
  • 1 measurement input
  • 1 measurement input
  • 1 measurement input
  • Outputs:
  • Mini USB-B
  • Mini USB-B
  • Mini USB-B
  • Mini USB-B
  • Mini USB-B
Specifications
Related Information
Electrodes
Product Information
Frequently Bought Together