Order Entry
Austria
Orders LinkContactUsLinkComponent
Sample cuvettes

Sample cuvettes

Sample cuvettes, 25 mm, 25 ml, Glass
Catalog # HACH2665908
Supplier:   Hach
Sample cuvettes
Catalog # HACH2665908
Supplier:  Hach
Sample cuvettes
Sample cuvettes

Sample cuvettes, 25 mm, 25 ml, Glass

Price
1 530,00 €
Log in for availability
1 * 8 ST
Product Details & Documents

Sample cuvettes with highly visible graduations.

 : For use with neoprene stopper – Not included.

Specifications
  • Volume:
    25 ml
  • Window material:
    Glass
  • Description:
    Sample cuvettes
  • Height:
    25 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.