Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid ≥96% for nitrogen analysis
Sulphuric acid ≥96% for nitrogen analysis
Supplier:  VWR Chemicals
Certificates
undefined
Sulphuric acid ≥96% for nitrogen analysis
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 96 %
  • Colouration:
    Max. 10 APHA
  • Ignition residue:
    Max. 2 ppm
  • Substances reducing KMnO4 (as SO2):
    Max. 2 ppm
  • Total N (Nitrogen):
    Max. 1 ppm

Specification Test Results

Frequently Bought Together