Order Entry
Austria
Orders LinkContactUsLinkComponent
Ammonia electrode storage solution

Ammonia electrode storage solution

Storage solution for ammonia ion selective electrodes, 500 mL
Catalog # HACH2541249
Supplier:   Hach
Ammonia electrode storage solution
Catalog # HACH2541249
Supplier:  Hach
CAS Number:  
Ammonia electrode storage solution
Ammonia electrode storage solution

Storage solution for ammonia ion selective electrodes, 500 mL

CAS Number:  
Price
70,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

For storage of ammonia ion selective electrodes.

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Storage solution for ammonia ion selective electrodes

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.