Orders LinkContactUsLinkComponent
Filter disc
Filter disc
3, 90 mm
Catalog # KAVA632424011309
Supplier:   Kavalier
Filter disc
Catalog # KAVA632424011309
Supplier:  Kavalier
Filter disc
Filter disc
3, 90 mm
Price
2 100,00 €
Log in for availability
1 * 20 ST
Product Details & Documents
Filter disc made from borosilicate glass. Sintered glass filter with ground edges.
Specifications
  • Ø:
    90 mm
  • Porosity:
    3
  • Seq:
    01