Order Entry
Austria
ContactUsLinkComponent
Universal pathogen DNA kit, Mag-Bind®
Universal pathogen DNA kit, Mag-Bind®
Supplier:  OMEGA BIO-TEK
Certificates
undefined
Universal pathogen DNA kit, Mag-Bind®
Supplier:  OMEGA BIO-TEK
Certificates

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Viral DNA
    Bacterial DNA
  • Sample type:
    Stool
    Biofluids (serum, plasma, saliva)
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Sample size:
    Variable
  • Description:
    Mag-Bind® Universal Pathogen DNA Kit

Specifications

Related Information

Frequently Bought Together