Chlorine reagent set
Supplier: Hach
Certificates
About this item
This chlorine reagent set has 100 tests each to test chlorine (free and total) by DPD colorimetic method.
Frequently Bought Together
This chlorine reagent set has 100 tests each to test chlorine (free and total) by DPD colorimetic method.