ContactUsLinkComponent
Hydrazine colour disc test kit, HY-2
Hydrazine colour disc test kit, HY-2
Catalog # HACH184900
Supplier:  Hach
CAS Number:  
undefined
Hydrazine colour disc test kit, HY-2
Catalog # HACH184900
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Hydrazine color disc test kit, HY-2
  • Tests per kit:
    300
  • Range:
    0 to 1 mg/l
Specifications
About this item
Proper maintenance and economy of operation of modern boilers and cooling towers require a careful balance between water quality parameters and treatment chemicals. This kit measures hydrazine, a common oxygen scavenger in boiler water, using the simple colour disc method.
Delivery: Includes two 100 ml bottles of deionised water, one reagent, two stoppers, one colour disc comparator, one colour disc, instructions, and a carrying case.