Orders LinkContactUsLinkComponent
Test kits, COD cell test, Spectroquant®, Supelco®
Test kits, COD cell test, Spectroquant®, Supelco®
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier:  Merck
Certificates
Test kits, COD cell test, Spectroquant®, Supelco®
11 Options Available Below
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*7 This method is officially recognised by the USEPA as an alternative method for the investigation of the following:
*7-1 wastewater
*7-2 drinking water
*7-3 respectively drinking water and wastewater
*8 COD cell tests fully comply with ISO 15705.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 1.14560.0001
  • 1.01796.0001
  • 1.14540.0001
  • 1.14895.0001
  • 1.14541.0001
  • 1.14690.0001
  • 1.14691.0001
  • 1.14555.0001
  • 1.01797.0001
  • 1.09772.0001
  • 1.09773.0001
  • M-Clarity quality level:
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
Specifications
Product Family Options
Product Information
  • Range
    Chemical method
    For unit
    Pk
    Availability
    Price
  • COD cell test*⁷⁻¹
    4,0 - 40,0 mg/L
    Chromosulphuric oxidation
    Prove, Nova 30/60
    25 Tests
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests - 174,00 €
    174,00 €
    1 * 25 Tests
       174,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.14560.0001
    M-Clarity quality level
    MQ100
  • COD cell test*⁸
    5,0 - 80,0 mg/L
    Chromosulphuric oxidation
    Prove, Nova 30/60, Multy, Move
    25 Tests
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests - 147,00 €
    147,00 €
    1 * 25 Tests
       147,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.01796.0001
    M-Clarity quality level
  • COD cell test*7-1
    10 - 150 mg/L
    Chromosulphuric oxidation
    Prove, Nova 30/60, Multy, Move, Picco
    25 Tests
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests - 174,00 €
    174,00 €
    1 * 25 Tests
       174,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.14540.0001
    M-Clarity quality level
    MQ100
  • COD cell test*⁷⁻¹
    15 - 300 mg/L
    Chromosulphuric oxidation
    Prove, Nova 30/60, Multy, Move
    25 Tests
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests - 133,00 €
    133,00 €
    1 * 25 Tests
       133,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.14895.0001
    M-Clarity quality level
    MQ100
  • COD cell test*⁷⁻¹
    25 - 1500 mg/L
    Chromosulphuric oxidation
    Prove, Nova 30/60, Multy, Move, Picco
    25 Tests
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests - 174,00 €
    174,00 €
    1 * 25 Tests
       174,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.14541.0001
    M-Clarity quality level
    MQ100
  • COD cell test*⁷⁻¹
    50 - 500 mg/L
    Chromosulphuric oxidation
    Prove, Nova 30/60, Multy, Move
    25 Tests
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests - 147,00 €
    147,00 €
    1 * 25 Tests
       147,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.14690.0001
    M-Clarity quality level
    MQ100
  • COD cell test*⁷⁻¹
    300 - 3500 mg/L
    Chromosulphuric oxidation
    Prove, Nova 30/60, Multy, Move
    25 Tests
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests - 133,00 €
    133,00 €
    1 * 25 Tests
       133,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.14691.0001
    M-Clarity quality level
    MQ100
  • COD cell test*⁷⁻¹
    500 - 10000 mg/L
    Chromosulphuric oxidation
    Prove, Nova 30/60, Multy, Move, Picco
    25 Tests
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests - 147,00 €
    147,00 €
    1 * 25 Tests
       147,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.14555.0001
    M-Clarity quality level
    MQ100
  • COD cell test*⁸
    5000 - 90000 mg/L
    Chromosulphuric oxidation
    Prove, Nova 30/60, Multy, Move
    25 Tests
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests
    Specifications:
    More
    Specifications
    Cat. No.
    1.01797.0001
    M-Clarity quality level
  • 1.09772.0001
    COD cell test (Hg-free)
    COD cell test (Hg-free)
    10 - 150 mg/L
    Chromosulphuric oxidation
    Prove, Nova 30/60, Move, Multy
    25 Tests
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests - 133,00 €
    133,00 €
    1 * 25 Tests
       133,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.09772.0001
    M-Clarity quality level
    MQ100
  • 1.09773.0001
    COD cell test (Hg-free)
    COD cell test (Hg-free)
    100 - 1500 mg/L
    Chromosulphuric oxidation
    Prove, Nova 30/60, Move, Multy
    25 Tests
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests - 133,00 €
    133,00 €
    1 * 25 Tests
       133,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.09773.0001
    M-Clarity quality level
    MQ100
COD Cell Test for seawater / high chloride contents
Product Information
  • Catalog #
    Pk
    Chemical method
    Description
    Range
    For unit
    Availability
    Price
  • 25 Tests
    Chromosulphuric oxidation
    COD cell test for seawater
    5,0 - 60,0 mg/L
    Prove, Nova 30/60, Multy, Move
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests - 232,00 €
    232,00 €
    1 * 25 Tests
       232,00 €
Product Information
  • Catalog #
    Pk
    Chemical method
    Description
    Range
    For unit
    Availability
    Price
  • 25 Tests
    Chromosulphuric oxidation
    COD cell test for seawater
    50 - 3000 mg/L
    Prove, Nova 30/60, Multy, Move
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests - 232,00 €
    232,00 €
    1 * 25 Tests
       232,00 €
Accessories
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Aborption cone for use with COD test for seawater 1.17059.0001
    Aborption cone for use with COD test for seawater 1.17059.0001
    1.15955.0001
    1
    Log in for availability
    1 * 1 ST - 228,00 €
Product Information
  • Catalog #
    Pack type
    Pk
    Availability
    Price
  • 1.06733.0501
    Plastic bottle
    500 g
    Log in for availability
    1 * 500 g