Jolting volumeter
Catalog # 614-0255
Supplier: GERBER FUNKE
Specifications
- Description:Jolting volumeter
- Pk:1
Specifications
About this item
This jolting volumeter is designed to determine the tamped volume or density of powdery and granular products.
Certifications: ISO 8967 and IDF 134: Milk powder. USP 616 method I: Pharmaceutical powder. ISO 8460: Instant coffee.