Order Entry
Austria
ContactUsLinkComponent
 
Dimethyl sulphoxide ≥99.9% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
  9224-01
 :  Avantor
   Safer Human and Environmental Health
 :  
undefined
Dimethyl sulphoxide ≥99.9% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
  9224-01
 :  Avantor
   Safer Human and Environmental Health
 :  

 

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    500 mL
  • Seq:
    2725
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

 

 

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
  • Appearance:
    Passes Test
  • Assay ((CH₃)₂SO) (by GC, corrected for water):
    ≥99.9 %
  • Residue after Evaporation:
    ≤0.0100 %
  • Titrable Acid (meq/g):
    ≤0.001
  • Water (by KF, coulometric):
    ≤0.1 %
  • Product Information (not specifications)::

 

 

 :
 :78.14 g/mol
 :13,03285
 :189 °C (1013 hPa)
 :18.5 °C
 :1.101 g/cm³ (20 °C)
 :87 °C
 :Ambient
 :200-664-3
 :67-68-5
 :(CH₃)₂SO
 :MFCD00002089