Order Entry
Austria
ContactUsLinkComponent
Guanidinium hydrochloride ≥98%, BAKER ANALYZED® for biochemistry, J.T.Baker®
Irritant
Guanidinium hydrochloride ≥98%, BAKER ANALYZED® for biochemistry, J.T.Baker®
Catalog # 4077-01
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Guanidinium hydrochloride ≥98%, BAKER ANALYZED® for biochemistry, J.T.Baker®
Catalog # 4077-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 98.0 %
  • Assay (NH₂C(:NH)NH₂·HCl):
    ≥98.0 %
  • Loss on Drying at 105°C:
    ≤0.3 %
  • UV Absorbance of 6M Solution - 275 nm, For Information Only:
    For Information Only
  • UV Absorbance of 6M Solution - 260 nm, For Information Only:
    175 - 188 °C
  • Melting Range(Lower):
    175 - 188 °C
  • Melting Range(Upper):
    175 - 188 °C

Specification Test Results

About this item

 :
 :95.53 g/mol
Merck Index:13,04578
Melting Pt:185 °C
Density:1.354 g/cm³ (20 °C)
EINECS:200-002-3
 :GHS07
Storage Temperature:Ambient
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026