Order Entry
Austria
ContactUsLinkComponent
Calcium carbonate ≥99.0% (dried basis, T), powder, BAKER ANALYZED® ACS, J.T.Baker®
Calcium carbonate ≥99.0% (dried basis, T), powder, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 1288-R
Supplier:  Avantor
CAS Number:  
undefined
Calcium carbonate ≥99.0% (dried basis, T), powder, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 1288-R
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Fiber drum
  • Pk:
    100 lb
  • Seq:
    7167

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • Assay (CaCO₃) (dried basis):
    ≥99.0 %
  • Insoluble in Dilute HCl:
    ≤0.01 %
  • Chloride (Cl):
    ≤0.001 %
  • Fluoride (F):
    ≤0.0015 %
  • Sulfate (SO₄):
    ≤0.01 %
  • Ammonium (NH₄):
    ≤0.003 %
  • Barium (Ba):
    ≤0.01 %
  • Heavy Metals (as Pb):
    ≤0.001 %
  • Iron (Fe):
    ≤0.003 %
  • Magnesium (Mg):
    ≤1.0 %
  • Potassium (K):
    ≤0.01 %
  • Sodium (Na):
    ≤0.1 %
  • Strontium (Sr):
    ≤0.1 %

Specification Test Results

About this item

 :
 :100.09 g/mol
Merck Index:13,01658
Melting Pt:825 °C
Density:2.71 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-439-9
CAS Number:471-34-1
Formula:CaCO₃
MDL Number:MFCD00010906