Order Entry
Austria
ContactUsLinkComponent
Trisodium phosphate dodecahydrate 98.0-102.0% (by acidimetry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Corrosive
Trisodium phosphate dodecahydrate 98.0-102.0% (by acidimetry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3836-07
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Trisodium phosphate dodecahydrate 98.0-102.0% (by acidimetry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 3836-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    98.0 - 102.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤2.5%
  • Assay (Na₃PO₄·12H₂O) (by acidimetry):
    98.0 - 102.0 %
  • Excess Alkali (as NaOH):
    ≤2.5 %
  • Insoluble Matter:
    ≤0.01 %
  • Chloride (Cl):
    ≤0.001 %
  • Sulfate (SO₄):
    ≤0.01 %
  • Heavy Metals (as Pb):
    ≤0.001 %
  • Trace Impurities - Iron (Fe):
    ≤0.001 %

Specification Test Results

About this item

 :
 :380.12 g/mol
Merck Index:13,08736
Melting Pt:∼75 °C
Density:1.62 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-509-8
UN:3262
ADR:8,III
REACH:01-2119489800-32-xxxx
 :GHS05
CAS Number:10101-89-0
Formula:Na₃PO₄·12H₂O
MDL Number:MFCD00149198