Lithium chloride ≥99%, granular, AR® ACS, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:7447-41-8
:
Formula:LiCl
MDL Number:MFCD00011078
Boiling Pt:1382
Melting Pt:614
Density:2.07 g/cm³ (20 °C)
Molecular Weight:42.39 g/mol
Merck Index:13,05550
EINECS:231-212-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:= 99 %
- ACS - Assay (LiCl):≥99 %
- Solubility:Passes Test
- ACS - Titrable Base (meq/g):≤0.008
- ACS - Calcium (Ca):≤0.01 %
- ACS - Heavy Metals (as Pb):≤0.002 %
- ACS - Insoluble Matter:≤0.01 %
- ACS - Iron (Fe):≤0.001 %
- ACS - Potassium (K):≤0.01 %
- ACS - Sodium (Na):≤0.20 %
- ACS - Sulfate (SO₄):≤0.01 %
- ACS - Barium (Ba):≤0.003 %
- ACS - Loss on Drying at 105°C:≤1.0 %
Specification Test Results
Frequently Bought Together