


Toluene ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:108-88-3
:
:C₆H₅CH₃
:MFCD00008512
:92.14 g/mol
:13,09607
:110.6 °C (1013 hPa)
:–95 °C
:0.867 g/cm³ (20 °C)
:4 °C
:Ambient
:203-625-9
:1294
:3,II
:05-2114615130-69
:GHS02
:GHS08
:GHS07
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:—
- Assay (C₆H₅CH₃) (by GC):≥99.5 %
- Color (APHA):≤10
- Residue after Evaporation:≤0.001 %
- Substances Darkened by H₂SO₄:Passes Test
- Sulfur Compounds (as S):≤0.003 %
- Water (by KF, coulometric):≤0.03 %
- Date Received: __ __ /__ __ /__ __:—
- Date Opened: __ __ /__ __ /__ __:—
- Meets Reagent Specifications for testing USP/NF monographs:—
Frequently Bought Together