Order Entry
Austria
ContactUsLinkComponent
L(+)-Tartaric acid ≥99.0%, granular, AR® ACS, Macron Fine Chemicals™
Corrosive
L(+)-Tartaric acid ≥99.0%, granular, AR® ACS, Macron Fine Chemicals™
Catalog # 2312-04
CAS Number:  
undefined
Corrosive
L(+)-Tartaric acid ≥99.0%, granular, AR® ACS, Macron Fine Chemicals™
Catalog # 2312-04
CAS Number:  

Specifications

  • Pack type:
    PE bottle, Mall Polystormor®
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    ≥99.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.001%
  • ACS - Assay:
    ≥99.0 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Iron (Fe):
    ≤5 ppm
  • ACS - Oxalate (C₂O₄):
    Passes Test
  • ACS - Phosphate (PO₄):
    ≤0.001 %
  • ACS - Residue after Ignition:
    ≤0.02 %
  • ACS - Sulfur Compounds (as S):
    ≤0.002 %

Specification Test Results

About this item

 :
Merck Index:13,09158
Boiling Pt:399 °C (1013 hPa)
Melting Pt:168…170 °C
Density:1.759 g/cm³ (20 °C)
Flash Pt:150 °C (closed cup)
 :150.09 g/mol
Storage Temperature:Ambient
EINECS:201-766-0
REACH:01-2119537204-47
 :GHS05
CAS Number:87-69-4
Formula:C₄H₆O₆
MDL Number:MFCD00064207