Order Entry
Austria
ContactUsLinkComponent
 
tri-Sodium citrate dihydrate 99.0-101.0% (by non aqueous titration), granular, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 
undefined
tri-Sodium citrate dihydrate 99.0-101.0% (by non aqueous titration), granular, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 

 

  • Meets ACS Reagent Chemical Requirements:
    99.0 - 101.0 %
  • For Laboratory,Research,or Manufacturing Use:
    99.0 - 101.0 %
  • ACS - Assay (C₆H₅Na₃O₇·2H₂O) (by non-aqueous titration):
    99.0 - 101.0 %
  • ACS - pH of 5% Solution at 25°C:
    7.0 - 9.0
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Chloride (Cl):
    ≤0.003 %
  • ACS - Sulfate (SO₄):
    ≤0.005 %
  • ACS - Ammonia (as NH₃):
    ≤0.003 %
  • ACS - Calcium (Ca):
    ≤0.005 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

 

Frequently Bought Together