Order Entry
Austria
ContactUsLinkComponent
 
 
 
2-Propanol ≥99.5%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
  3043-25
 :  
undefined
 
 
2-Propanol ≥99.5%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
  3043-25
 :  

 

  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • For Laboratory,Research,or Manufacturing Use:
    Min. 99.5 %
  • ACS - Assay (CH₃CHOHCH₃):
    ≥99.5 %
  • Carbonyl Compounds - Propionaldehyde:
    ≤0.002 %
  • Carbonyl Compounds - Acetone:
    ≤0.002 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤0.0005 %
  • Solubility in H₂O:
    Passes Test
  • Titrable Acid or Base (meq/g):
    ≤0.0001
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.05 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 205 nm:
    ≤1.00
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:
    ≤1.00
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:
    ≤0.30
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm:
    ≤0.15
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 245 nm:
    ≤0.08
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm:
    ≤0.02
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm:
    ≤0.04
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm:
    ≤0.03
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm:
    ≤0.02
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 - 330 nm:
    ≤0.01

 

 

 :
 :60.1 g/mol
 :13,05228
 :82 °C (1013 hPa)
 :–89 °C
 :0.786 g/cm³ (20 °C)
 :12 °C
 :Ambient
 :200-661-7
 :1219
 :3,II
 :01-2119457558-25
 :GHS02
 :GHS07
 :67-63-0
 :(CH₃)₂CHOH
 :MFCD00011674