Order Entry
Austria
ContactUsLinkComponent
Sodium acetate, anhydrous ≥99%, purified, J.T.Baker®
Sodium acetate, anhydrous ≥99%, purified, J.T.Baker®
Catalog # 3472-05
Supplier:  Avantor
CAS Number:  
undefined
Sodium acetate, anhydrous ≥99%, purified, J.T.Baker®
Catalog # 3472-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    2 kg
  • Seq:
    4058

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • Assay (CH₃COONa):
    ≥99.0 %
  • Insoluble Matter:
    ≤0.01 %
  • pH of 5% Solution at 25°C:
    7.0 - 9.2
  • Heavy Metals (as Pb):
    ≤0.002 %
  • Loss on Drying at 120°C:
    ≤1.0 %

Specification Test Results

About this item

 :
 :82.03 g/mol
Merck Index:12,08711
Boiling Pt:881.4 °C (1013 hPa)
Melting Pt:324 °C
Density:1.528 g/cm³ (20 °C)
Flash Pt:>250 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-823-8
REACH:01-2119485123-42
CAS Number:127-09-3
Formula:H₃CCOONa
MDL Number:MFCD00012459