Tubings, insulated
Supplier: LAUDA
Certificates
About this item
These metal hoses with thermal insulation consist of flexible corrugated stainless steel tubing and are unaffected by the usual bath liquids. The hose is secured to the connection with a ring nut. The fitting consists of a taper-cone system and not involves any flexible seal.
Frequently Bought Together