Bulb pipettes, Class A, SP Wilmad-LabGlass
WILMLG-9350-136
- Colour code:Blue
- Capacity:200 ml
- Seq:02
- Pk:1
Glass.
- Calibrated to deliver at 20 °C
- Fabricated to meet the requirements of accuracy, appearance, glass quality and calibration line
- Colour coded
: ASTM E-969.