Order Entry
Austria
ContactUsLinkComponent
NMR tube carrier, SP Wilmad-LabGlass
NMR tube carrier, SP Wilmad-LabGlass
Catalog # WILMWG-6192
Supplier:  SP Industries
undefined
NMR tube carrier, SP Wilmad-LabGlass
Catalog # WILMWG-6192
Supplier:  SP Industries

Specifications

  • Description:
    Tube carrier for 5 mm O-Ø NMR tubes
  • Pk:
    3

Specifications

About this item

Made from unbreakable polymer material with a snap-on cap, these tube carriers feature a shirt clip that keeps it secured in lab coat pockets.

  • Protection is provided for the sample and lab personnel during transport
  • Comes in a pack of three white NMR tube carriers
  • Holds 5×177,8 mm NMR tubes