Cell spreaders, L-shaped
Artikelnr. HEATHS8171B
Supplier: HEATHROW
Specifications
- Packed:10/bag
- Description:Cell spreaders, L-shaped, disposable, sterile
- Length:146 mm
- Pk:500
Specifications
About this item
ABS disposable L-shaped cell spreader with smooth surface that will not tear agar plate or disturb media layer. Supplied sterile for immediate use without flaming.