Immersion thermostats, IC series
: IKA
The IC immersion circulators have an extended temperature range for tempering fluids in open and covered water baths up to 200/250 °C. The LED/ graphic TFT Display illustrates all relevant process parameters. Units feature push and turn buttons for intuitive menu navigation. IC immersion circulators ensure short heating up times and have a temperature stability of ±0,01/0,02 K (basic/control model). Both models feature optical and acoustic safety warnings, an adjustable safety circuit, a powerful pressure/suction pump and can be used for internal and external applications.
- A Wireless Controller (WiCo) enables safe operation of control models from a distance of up to 10 meters, e.g. in front of a closed fume hood (control device only)
- Large easy-to-read LED display for pump speed and temperature on basic units, control units have a TFT display for temperature, pump speed as well as liquid level and safety temperature
- The control models have timer function: 0 to 99 h 59 min, and a Mulit IO (Input/Output) interface to connect and control for e.g. electronic magnetic valves. Cooling coil is already included
- USB/RS232 interface for PC control, documentation using labworldsoft® and for online firmware updates, as well as PT100 interface for external temperature control
- The user can switch from internal to external temperature regulation with only one key using the control model
- Integrated connections M16×1 for external applications
Max. flow rate/max pressure:
31 L/min, 610 mbar, (pressure), 450 mbar (suction)
* Lower working temperatures possible with connection to a chiller.
: Comply with DIN 12876, Safety class III, (FL) for use with flammable liquids. Units are designed in accordance to UL 61010-1.
: Supplied with telescopic stainless steel bath-bridge for connection to water baths with different volumes. Both devices have a PT100 interface for external temperature control.
Frequently Bought Together