Order Entry
Austria
ContactUsLinkComponent
Glial cell line-derived neurotrophic factor (GDNF) Rapid™ ELISA kit: rat
Glial cell line-derived neurotrophic factor (GDNF) Rapid™ ELISA kit: rat
Supplier:  Biosensis
Certificates
undefined
Glial cell line-derived neurotrophic factor (GDNF) Rapid™ ELISA kit: rat
Supplier:  Biosensis

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Rat
  • Target protein:
    GDNF
  • Cross reactivity:
    No cross-reactivity was observed for the following proteins tested at 25 ng/mL: brain-derived neurotrophic factor (rhBDNF), nerve growth factor (rhNGF), neurotrophin-3 (rhNT-3), rhNT-4/5, vascular endothelial growth factor (rhVEGF), recombinant human Neurturin, Artemin and Persephin.
  • Specificity:
    Rat. The antibodies used in this kit detect mouse and human GDNF.
  • Detection range:
    7,8 – 500 pg/ml
  • Sensitivity:
    Typical limit of detection (LOD) for rrGDNF is <15 pg/ml determined as blank value plus 3× standard deviation of blank OD (n=10).

Specifications

Frequently Bought Together