Order Entry
Austria
ContactUsLinkComponent
Nerve growth factor Receptor (NGFR/P75ECD) Rapid™ ELISA kit: Mouse
Nerve growth factor Receptor (NGFR/P75ECD) Rapid™ ELISA kit: Mouse
Supplier:  Biosensis
Certificates
undefined
Nerve growth factor Receptor (NGFR/P75ECD) Rapid™ ELISA kit: Mouse
Supplier:  Biosensis

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Cross reactivity:
    The antibodies used in this ELISA kit does not cross-react with human p75ECD protein
  • Detection range:
    62,5 – 4000 pg/ml
  • Sensitivity:
    Typically detects down to 20 to 40 pg/ml of mouse p75ECD (defined as blank OD plus 3x the standard deviation of the blank OD, n=10)

Specifications

Frequently Bought Together