Order Entry
Austria
ContactUsLinkComponent
Neurotrophin 3 (NT3) Rapid™ ELISA kit
Neurotrophin 3 (NT3) Rapid™ ELISA kit
Supplier:  Biosensis
Certificates
undefined
Neurotrophin 3 (NT3) Rapid™ ELISA kit
Supplier:  Biosensis

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
    Rat
    Mouse
  • Target protein:
    NT3
  • Specificity:
    Human. The capture and detection antibodies used detect NT3 from other species including mouse, rat and monkey, thus it is expected that this NT3 ELISA will also react with those species.
  • Detection range:
    15,6 – 1000 pg/ml
  • Sensitivity:
    Typically detects a minimum of 5 pg/ml NT3 (defined as 150% of blank value).

Specifications

Frequently Bought Together