ContactUsLinkComponent
Cleanroom drop-down overalls, BioClean-D™
Cleanroom drop-down overalls, BioClean-D™
Supplier:  Nitritex
Certificates
undefined
Cleanroom drop-down overalls, BioClean-D™
Supplier:  Nitritex
Certificates
Specifications
  • Catalog No:
  • 112-4987
  • 112-4984
  • 112-4982
  • 112-4981
  • 112-4985
  • 112-4986
  • 112-4983
  • 112-4980
  • Model:
  • Drop-down
  • Drop-down
  • Drop-down
  • Drop-down
  • Drop-down
  • Drop-down
  • Drop-down
  • Drop-down
  • Cleanroom class:
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • Garment application:
  • Cleanroom, Laboratory
  • Cleanroom, Laboratory
  • Cleanroom, Laboratory
  • Cleanroom, Laboratory
  • Cleanroom, Laboratory
  • Cleanroom, Laboratory
  • Cleanroom, Laboratory
  • Cleanroom, Laboratory