Order Entry
Austria
ContactUsLinkComponent
Ion exchange chromatography media, Capto™ S ImpAct
Ion exchange chromatography media, Capto™ S ImpAct
Supplier:  Cytiva
Certificates
undefined
Ion exchange chromatography media, Capto™ S ImpAct
Supplier:  Cytiva

Specifications

  • pH range:
    4 - 12
  • Matrix:
    High-flow agarose
  • Ligand:
    -SO3-
  • Storage temperature:
    4...30 °C
  • Binding capacity:
    >85 mg BSA/ml, >90 mg lysozyme/ml, >100 mg IgG/ml
  • Total ionic capacity:
    37 - 63 umol (H+)/ml

Specifications

Related Information

Frequently Bought Together