Order Entry
Austria
ContactUsLinkComponent
Capto™ MMC ImpRes multimodal chromatography media, Cytiva
Capto™ MMC ImpRes multimodal chromatography media, Cytiva
Supplier:  Cytiva
Certificates
undefined
Capto™ MMC ImpRes multimodal chromatography media, Cytiva
Supplier:  Cytiva

Specifications

  • pH range:
    3 - 12
  • Matrix:
    High-flow agarose
  • Storage temperature:
    4 - 30 °C
  • Binding capacity:
    60 - 90 mg Mab/ml

Specifications

Frequently Bought Together