TISAB solution for Fluoride Determination, Supelco®
Catalog # 1.15368.0500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 mL
- Description:TISAB solution for fluoride determination
- M-Clarity quality level:MQ100
- pH:5,0 - 6,0
- Contents:NaCl, Titriplex IV