Inverted microscope, CKX53
About this item
Ergonomically designed and enhanced imaging for cell culture, the CKX53 is designed for cell sampling and manipulation, especially for fluorescence observations.
- Observations in lightfield transmission and phase contrast evolutionary to fluorescence
- High contrast thanks to integrated phase system (IPC) (without ring change)
- Three-position fluorescence illuminator: Two sets of filters for excitation (blue and green), empty position, manual shutter and neutral filter slide 6%
Compatible with CKX3-SLP and IX2-SL condenser slides, CKX3-SPLIC, CKX3-SLPAS, IX2-SPLC, IX2-SLPH1, IX2-SLPH2 optical elements, CKX3-MVR object guide, CKX3-HO35DM Petri dish rack, reflective fluorescence illuminator and mercury, metal halide or LED light sources.
: Included CACHN 10× / 0,25 Ph / IPC lens, power cable, platinum insert, Allen key, power supply, two camera cable clips with screws - contrast-inverted 3-D views, optional.
3 Options Available Below
Product Details & Documents
Ergonomically designed and enhanced imaging for cell culture, the CKX53 is designed for cell sampling and manipulation, especially for fluorescence observations.
- Observations in lightfield transmission and phase contrast evolutionary to fluorescence
- High contrast thanks to integrated phase system (IPC) (without ring change)
- Three-position fluorescence illuminator: Two sets of filters for excitation (blue and green), empty position, manual shutter and neutral filter slide 6%
Compatible with CKX3-SLP and IX2-SL condenser slides, CKX3-SPLIC, CKX3-SLPAS, IX2-SPLC, IX2-SLPH1, IX2-SLPH2 optical elements, CKX3-MVR object guide, CKX3-HO35DM Petri dish rack, reflective fluorescence illuminator and mercury, metal halide or LED light sources.
: Included CACHN 10× / 0,25 Ph / IPC lens, power cable, platinum insert, Allen key, power supply, two camera cable clips with screws - contrast-inverted 3-D views, optional.
Product Family Options
Fluorescence equipment
Product Information
- AvailabilityPrice
- OLYMN2757800Lamp housing with lampholder and aspherical lens for mercury vapor lamp 100 WLamp housing with lampholder and aspherical lens for mercury vapor lamp 100 WLog in for availability1 * 1 ST
- OLYMN2712500Single-band fluorescence cube, broadband blue excitation (460 - 495 nm), low-pass filter, beam splitter: 505 mm, interference filter: 510 nmSingle-band fluorescence cube, broadband blue excitation (460 - 495 nm), low-pass filter, beam splitter: 505 mm, interference filter: 510 nmLog in for availability1 * 1 ST
- OLYMN5228400Reflected fluorescence illuminator
Objectives
Product Information
- Catalog #MagnificationDescriptionAvailabilityPrice
Accessories
Product Information
- Catalog #AvailabilityPrice
- Dust Cover for CKX Microscopes
Product Information
- Catalog #AvailabilityPrice
- Power cable with elbow
Product Information
- Catalog #AvailabilityPrice
- CKX3-SLP phase contrast slider, pre-centered for CKX53 and IX53CKX3-SLP phase contrast slider, pre-centered for CKX53 and IX53OLYMN5228500Log in for availability1 * 1 ST - 114,00 €