Orders LinkContactUsLinkComponent
Adapter kit
Adapter kit
Adaptor kit (4 mm male / 4 mm female)
Catalog # PSA410
Supplier:   Cytiva
Adapter kit
Catalog # PSA410
Supplier:  Cytiva
Adapter kit
Adapter kit
Adaptor kit (4 mm male / 4 mm female)
Price
46,80 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Connects 4 mm output jack to 4 mm high voltage leads.
Note that this adaptor kit is not CE compliant.
Ordering information: Supplied with Allen wrench.
Specifications
  • Description:
    Adaptor kit (4 mm male / 4 mm female)