Methyl dodecylbenzene sulphonate ≥99% (by titrimetric analysis) for surfactant analysis, Sigma-Aldrich®
Catalog # 1.12148.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (two-phase titration):≥99
- Identity (IR-spectrum):Passes test
- Free acid (as dodecyl benzene sulfonic acid):≤0,5%
- Density:1,03 g/cm3
- Solubility:(20 °C) slightly soluble
- Storage:No restrictions
Specification Test Results
About this item
Synonyms: Benzenesulphonic acid,dodecyl-, methyl ester, methyl 2-dodecylbenzenesulphonate, Dodecylbenzenesulphonic acid methyl ester
:
:340.53 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:257-847-6
CAS Number:52319-06-9
Formula:C₁₉H₃₂O₃S
MDL Number:MFCD00078710