ContactUsLinkComponent
Standards for spectrophotometer calibration, Supelco®
Standards for spectrophotometer calibration, Supelco®
Catalog # 1.08159.0100
Supplier:  Merck
CAS Number:  
Standards for spectrophotometer calibration, Supelco®
Catalog # 1.08159.0100
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    Holmium perchlorate, density 1,136 g/cm3, soluble at 20 °C, with absorbance of 241,1±0,4 nm
  • M-Clarity quality level:
    MQ100
Specifications
About this item
Standard solutions are used for the calibration of spectrophotometers.