Electronic comparators, EComparator series
Lieferant: Lovibond Tintometer
Zertifikate
Über diesen Artikel
The Lovibond® EComparator series provides an easy way to transition from subjective visual measurement to a non subjective, accurate electronic measurement.
- Digital, portable, push-button technology
- On-screen colour
- On-screen numerical display
- User friendly ergonomics and intuitive interface guarantee new users can be quickly trained and easily supported
- Large data storage (>20000 readings)
- USB connectivity ensures readings can be stored and shared easily and quickly
Delivery: Instrument supplied with USB cable, USB power supply, 4× AA batteries, 2× liquid reference solutions, glass reference standards, cell divider, glass and plastic cells, mini screwdriver all contained in a plastic carrying case.
Spezifikationen
- Catalog No:
- TINT162010
- TINT163010
- TINT162020
- TINT163020
- Model:
- EC 2000 Pt Co
- EC 3000 ASTM Color
- EC 2000 Gardner
- EC 3000 Saybolt
- Parameters:
- Pt-Co
- ASTM Color
- Gardner colour scale
- Saybolt
- Range:
- 0 - 500
- 0 - 8
- 0 - 18
- −16...+30
- Resolution:
- 1 Pt-Co unit
- 0,1
- 0,1 Gardner unit
- 1 Saybolt unit
- Repeatability:
- ±3% +1 Pt-Co unit
- ±0,1
- ±0,2
- ±1
- Light source:
- White LED
- White LED
- White LED
- White LED
- Path length:
- 50 mm
- 33 mm
- 10 mm
- 50 mm
- Comparator view:
- 2 fields
- 3 fields
- 2 fields
- 3 fields
- Standard conformances:
- ASTM D1209
- ASTM D1500, D6045
- ASTM D1544, D6166
- ASTM D156, D6045
- Power supply:
- USB or 4×AA batteries
- USB or 4×AA batteries
- USB or 4×AA batteries
- USB or 4×AA batteries
- W×D×H:
- 106×210×57 mm
- 106×210×57 mm
- 106×210×57 mm
- 106×210×57 mm
Spezifikationen
Frequently Bought Together