ContactUsLinkComponent
VWR®, Disposable particle filtering respirators, N95
  VWRU89201-508
undefined
VWR®, Disposable particle filtering respirators, N95
  VWRU89201-508
  • Type:
    N-95
  • Seq:
    01
  • Size:
    M/L
  • Pk:
    20
  • With valve:
    -
The disposable lightweight N95 respirators combine excellent safety and comfort features to ensure a secure fit and a reliable protection.
  • Protection against solid and liquid particles that do not contain oil
  • Lightweight construction allows comfortable, extended use
  • An adjustable noseclip provides a secure seal against the user's face
 : NIOSH