VWR®, Bench Protectors with Leakproof Moisture Barrier
Catalog # VWRU89126-792
Supplier: VWR Collection
Specifications
- Description:Roll
- L×W:508×915000 mm
- Seq:03
- Environmentally Preferable:
- Pk:2
Specifications
About this item
These super absorbent bench protectors feature a two-layer, cushioned construction for use in a laboratory or veterinary setting. The air-lay cellulose layer provides cushion for laboratory equipment and glassware to prevent breakage, while the leakproof plastic backsheet protects tables and benches from spills and moisture.
- Two layers of protection and leakproof backsheet
- Available as rolls or mats
- Can be cut to desired size