Latex gloves, Rotiprotect®
Supplier: Roth Carl
Certificates
About this item
These disposable gloves are suitable for protection against complex risks and for food processing.
- Micro coarse outer surface
- Smooth inner surface, lightly powdered
- Very smooth and pleasantly soft
- Thin rolled edge
- Thiuram-free
Certifications: EN 374; Cat. III; AQL 1,5
Specifications
- Catalog No:
- 112-0044
- 112-0043
- 112-0045
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex
- Latex
- Latex
- Powdered / powder-free:
- Low-powdered
- Low-powdered
- Low-powdered
- Finish:
- Micro-textured
- Micro-textured
- Micro-textured
- Cuff style:
- Rolled
- Rolled
- Rolled
- EN norm:
- EN 374
- EN 374
- EN 374