Biohazard Waste Can Liner Bags
JUSU05901
- Ø×H:838×610 mm
- Capacity:56,8 L
- Colour:Red
- Description:Biohazard waste can liner bags
- Thickness:11 µm
- Pk:100
For safe, temporary collection of biological waste or contaminated materials. Red color is easily identifiable as biohazard bags.
- 15 Gallon (56,8 L) capacity bag fits Justrite Biohazard cans of 6 and 10 Gallon sizes
- High density PE of 11 microns thickness provides secure handling to avoid punctures and tears
- Tubular construction with a single side seal and star seal bottom provide a stable, leak proof bag
- Specifically labeled for Biohazard material handling
Printing includes English, Spanish and French translation along with the international recognizable biohazard symbol.