Orders LinkContactUsLinkComponent
Drum faucets
Drum faucets
Drum faucets
Drum faucets
These faucets can be simply placed on your drum for leakproof dispensing of flammable liquids. The brass construction with a polytetrafluoroethylene (PTFE) seal ensures excellent chemical resistance.
  • Safe and easy dispensing
  • FM approved
The faucets feature an internal flame arrester that prevents flashback of fire into the drum for added safety.
Product Family Options
  • Description
    Thread
  • Internal flame arrester, self-closing, wetted PTFE O-rings
    3/4"
    1 * 1 ST
    1 * 1 ST - 73,60 €
    73,60 €
    1 * 1 ST
       73,60 €
  • Internal flame arrester, adjustable
    3/4"
    1 * 1 ST
    1 * 1 ST - 78,10 €
    78,10 €
    1 * 1 ST
       78,10 €